Assay Details
| Gene Symbol: Zc3hav1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172467.3 | NP_766055.1 | 4 | 92 | ||
| GenBank mRNA | AK088451.1 | - | 1 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zc3hav1l
Genomic Map