Assay Details
| Gene Symbol: Gm16532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029648.1 | - | 217 | 120 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell division cycle 37 | Cdc37 |
p50, p50Cdc37 |
-2.6kb | Sense | Chr. 9 - 21138541 - 21149977 on NCBI Build GRCm38 |
| Downstream |
phosphodiesterase 4A, cAMP specific |
Pde4a |
D9Ertd60e, Dpde2 |
13.0kb | Anti-sense | Chr. 9 - 21165683 - 21213248 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm16532
Genomic Map