| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900011588.mm.1 |
NM_175013.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011588.mm.1 |
NM_175013.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011588.mm.1 |
TC1900001286.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011585.mm.1 |
NM_175013.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011585.mm.1 |
NM_175013.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011585.mm.1 |
TC1900001286.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011586.mm.1 |
NM_175013.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011586.mm.1 |
NM_175013.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011586.mm.1 |
TC1900001286.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011587.mm.1 |
NM_175013.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011587.mm.1 |
NM_175013.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011587.mm.1 |
TC1900001286.mm.1 |
MTA TC |