Assay Details
| Gene Symbol: Itlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007552.1 | NP_001007553.1 | 6 - 7 | 754 | 145 | |
| GenBank mRNA | AY217760.1 | - | 6 - 7 | 754 | 145 | |
More Information
Set Membership: |
|

Itlnb
Genomic Map