Assay Details
| Gene Symbol: Rps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009096.3 | NP_033122.1 | 2 - 3 | 176 | 159 | |
| GenBank mRNA | AK012284.1 | - | 1 - 2 | 146 | 159 | |
| AK012828.1 | - | 2 - 3 | 177 | 159 | ||
| AK013333.1 | - | 1 - 2 | 147 | 159 | ||
| AK013825.1 | - | 176 | 158 | |||
| AK028205.1 | - | 2 - 3 | 179 | 159 | ||
| AK050609.1 | - | 2 - 3 | 177 | 159 | ||
| AK146952.1 | - | 2 - 3 | 177 | 159 | ||
| AK161991.1 | - | 2 - 3 | 179 | 159 | ||
| AK166727.1 | - | 2 - 3 | 222 | 159 | ||
| AK168050.1 | - | 2 - 3 | 177 | 159 | ||
| AK168496.1 | - | 2 - 3 | 156 | 159 | ||
| AK169624.1 | - | 2 - 3 | 175 | 158 | ||
| BC090392.1 | - | 2 - 3 | 163 | 4459055 | 159 | |
| BC092050.1 | - | 2 - 3 | 163 | 3666815 | 159 | |
| Y00348.1 | - | 2 - 3 | 166 | 159 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps6
Genomic Map