Assay Details
| Gene Symbol: Ap3s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF084575.1 | - | 3 - 4 | 353 | 142 | |
| AK002565.1 | - | 3 - 4 | 379 | 142 | ||
| AK137489.1 | - | 3 - 4 | 374 | 142 | ||
| AK152100.1 | - | 3 - 4 | 385 | 142 | ||
| AK194047.1 | - | 1 - 2 | 135 | 142 | ||
| BC012656.1 | - | 3 - 4 | 364 | 4187607 | 142 | |
| BC108327.1 | - | 3 - 4 | 372 | 30713247 | 142 | |
| BC131985.1 | - | 3 - 4 | 354 | 40130262 | 142 | |
| BC145234.1 | - | 3 - 4 | 354 | 142 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ap3s1
Genomic Map