Assay Details
| Gene Symbol: Myl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010860.3 | NP_034990.1 | 3 - 4 | 183 | 176 | |
| GenBank mRNA | AK143885.1 | - | 3 - 4 | 213 | 176 | |
| AK167106.1 | - | 3 - 4 | 213 | 176 | ||
| AK167191.1 | - | 3 - 4 | 219 | 176 | ||
| AK167859.1 | - | 3 - 4 | 213 | 176 | ||
| AK168450.1 | - | 3 - 4 | 234 | 176 | ||
| AK168869.1 | - | 3 - 4 | 215 | 176 | ||
| BC026760.1 | - | 3 - 4 | 183 | 3990090 | 176 | |
| BC081470.1 | - | 3 - 4 | 200 | 6824167 | 176 | |
| BC089485.1 | - | 3 - 4 | 206 | 30277988 | 176 | |
| BC092214.1 | - | 3 - 4 | 191 | 5684607 | 176 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096257 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027969 | BC026760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027969 | NM_010860.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587346 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170222 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096257 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027969 | BC026760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027969 | NM_010860.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243727 | BC026760.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243727 | BC081470.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243727 | BC089485.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243727 | BC092214.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243727 | AK143885.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243726 | BC026760.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243726 | NM_010860.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541680 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170222 | - | - |
| Whole Mouse Genome G4122A | A_51_P243727 | BC026760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243727 | BC081470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243727 | BC089485.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243727 | BC092214.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243727 | AK143885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243726 | BC026760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243726 | NM_010860.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132235 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170222 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08636022 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08636033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08636050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08636052 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10503400 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10420012 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435041_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10420011 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581308 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10503399 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592847 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373396 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10354372 | - | - |
| Mouse Genome 430 2.0 Array | 1424269_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435041_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435041_at | - | - |
| Mouse Genome 430 2.0 Array | 1434396_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434396_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424269_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023595.mm.1 | NM_010860.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023595.mm.1 | TC1000003145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023599.mm.1 | NM_010860.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023599.mm.1 | TC1000003145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023601.mm.1 | NM_010860.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023601.mm.1 | TC1000003145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023600.mm.1 | NM_010860.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023600.mm.1 | TC1000003145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023602.mm.1 | NM_010860.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023602.mm.1 | TC1000003145.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435041_at | - | - |
| Mouse Expression Set 430 | 1435041_PM_at | - | - |
| Mouse Expression Set 430 | 1434396_a_at | - | - |
| Mouse Expression Set 430 | 1434396_PM_a_at | - | - |
| Mouse Expression Set 430 | 1424269_a_at | - | - |
| Mouse Expression Set 430 | 1424269_PM_a_at | - | - |
| Murine Genome U74v2 Set | 97542_PM_at | - | - |
| Murine Genome U74v2 Set | 97542_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424269_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434396_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435041_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435041_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424269_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434396_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 893658 | - | - |
| ABI Mouse Genome Survey Microarray | 893890 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 60563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 60563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6100152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000060563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006100152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775661 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775661 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003895.1_38-S | - | - |
| mouse-6 v1.1 (Illumina) | scl017904.3_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 60563 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6100152 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017904.3_16-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003895.1_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6100152 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 60563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2900910 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2900910 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243433 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775660 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017904.3_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003895.1_38-S | - | - |

Myl6
Genomic Map