Assay Details
| Gene Symbol: Actr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205385.1 | NP_001192314.1 | 9 - 10 | 1066 | 151 | |
| NM_001205386.1 | NP_001192315.1 | 9 - 10 | 1063 | 151 | ||
| NM_023735.2 | NP_076224.1 | 8 - 9 | 1258 | 151 | ||
| GenBank mRNA | AK004554.1 | - | 8 - 9 | 1072 | 151 | |
| AK083343.1 | - | 8 - 9 | 1076 | 151 | ||
| AK145536.1 | - | 8 - 9 | 1078 | 151 | ||
| AK150487.1 | - | 8 - 9 | 960 | 151 | ||
| AK150589.1 | - | 8 - 9 | 1036 | 151 | ||
| AK150991.1 | - | 8 - 9 | 1016 | 151 | ||
| AK152865.1 | - | 8 - 9 | 1053 | 151 | ||
| AK153079.1 | - | 8 - 9 | 1079 | 151 | ||
| AK153150.1 | - | 8 - 9 | 1053 | 151 | ||
| AK153204.1 | - | 8 - 9 | 959 | 151 | ||
| AK159135.1 | - | 8 - 9 | 1045 | 151 | ||
| AK159668.1 | - | 8 - 9 | 970 | 151 | ||
| AK159775.1 | - | 8 - 9 | 1057 | 151 | ||
| AK160047.1 | - | 8 - 9 | 918 | 151 | ||
| AK166529.1 | - | 9 - 10 | 1010 | 151 | ||
| AK166586.1 | - | 9 - 10 | 1010 | 151 | ||
| AK167148.1 | - | 8 - 9 | 1017 | 151 | ||
| AK167891.1 | - | 8 - 9 | 1015 | 151 | ||
| AK168568.1 | - | 8 - 9 | 945 | 151 | ||
| AK168771.1 | - | 8 - 9 | 1031 | 151 | ||
| AK197794.1 | - | 1 - 2 | 89 | 151 | ||
| AK208071.1 | - | 1 - 2 | 89 | 151 | ||
| AK216313.1 | - | 1 - 2 | 89 | 151 | ||
| BC005557.1 | - | 8 - 9 | 1021 | 3156748 | 151 | |
| BC080806.1 | - | 9 - 10 | 1024 | 5350752 | 151 | |
| BC108386.1 | - | 8 - 9 | 989 | 5051125 | 151 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Actr3
Genomic Map