Assay Details
| Gene Symbol: Eef1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026007.4 | NP_080283.3 | 3 - 4 | 404 | 159 | |
| GenBank mRNA | AF321126.1 | - | 2 - 3 | 247 | 159 | |
| AK007869.1 | - | 3 - 4 | 249 | 159 | ||
| AK007991.1 | - | 3 - 4 | 252 | 158 | ||
| AK011951.1 | - | 2 - 3 | 246 | 159 | ||
| AK011973.1 | - | 3 - 4 | 251 | 159 | ||
| AK012431.1 | - | 3 - 4 | 252 | 157 | ||
| AK014277.1 | - | 3 - 4 | 251 | 159 | ||
| AK050636.1 | - | 3 - 4 | 255 | 159 | ||
| AK081859.1 | - | 3 - 4 | 252 | 159 | ||
| AK145519.1 | - | 3 - 4 | 405 | 159 | ||
| AK145565.1 | - | 3 - 4 | 253 | 159 | ||
| AK147980.1 | - | 3 - 4 | 252 | 159 | ||
| AK159644.1 | - | 3 - 4 | 251 | 159 | ||
| AK159768.1 | - | 3 - 4 | 251 | 159 | ||
| AK166765.1 | - | 3 - 4 | 250 | 159 | ||
| AK168749.1 | - | 3 - 4 | 251 | 159 | ||
| BC023495.1 | - | 2 - 3 | 214 | 5251765 | 159 | |
| BC083071.1 | - | 3 - 4 | 236 | 6329652 | 159 | |
| BC099413.1 | - | 2 - 3 | 232 | 30839877 | 159 | |
| BC117547.1 | - | 2 - 3 | 247 | 40105371 | 159 | |
| BC118522.1 | - | 2 - 3 | 247 | 40105372 | 159 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023495.1 transcripts.
Set Membership: |
|

Eef1g
Genomic Map