Assay Details
Gene Symbol: Ren1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031192.3 | NP_112469.1 | 5 - 6 | 718 | 143 | |
GenBank mRNA | AK052685.1 | - | 5 - 6 | 718 | 143 | |
AK085309.1 | - | 5 - 6 | 718 | 143 | ||
BC061053.1 | - | 5 - 6 | 716 | 30313659 | 143 | |
X16642.1 | - | 5 - 6 | 700 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
