Assay Details
Gene Symbol: Ren1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031192.3 | NP_112469.1 | 6 - 7 | 839 | 107 | |
GenBank mRNA | AK052685.1 | - | 6 - 7 | 839 | 107 | |
AK085309.1 | - | 6 - 7 | 839 | 107 | ||
BC024331.1 | - | 6 - 7 | 808 | 107 | ||
BC061053.1 | - | 6 - 7 | 837 | 30313659 | 107 | |
X16642.1 | - | 6 - 7 | 821 | 107 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
