Assay Details
| Gene Symbol: Fam89b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181452.2 | NP_852117.2 | 1 - 2 | 427 | 122 | |
| GenBank mRNA | AB012726.1 | - | 1 - 2 | 412 | 122 | |
| AF212920.1 | - | 1 - 2 | 409 | 122 | ||
| BC030163.1 | - | 1 - 2 | 398 | 3674980 | 122 | |
| BM942073.1 | - | 1 - 2 | 353 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam89b
Genomic Map