Assay Details
Gene Symbol: Prl2c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181852.1 | NP_862900.1 | 3 - 4 | 321 | 73 | |
GenBank mRNA | AF128884.1 | - | 3 - 4 | 321 | 73 | |
AK145824.1 | - | 4 - 5 | 516 | 73 | ||
BC132514.1 | - | 3 - 4 | 353 | 40130791 | 73 | |
BC132516.1 | - | 3 - 4 | 353 | 40130793 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
