Assay Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012027.2 | NP_036157.2 | 18 - 19 | 2918 | 164 | |
| NM_201245.3 | NP_957697.1 | 18 - 19 | 2918 | 164 | ||
| GenBank mRNA | AB093269.1 | - | 6 - 7 | 4743 | 164 | |
| AK035435.1 | - | 12 - 13 | 2208 | 164 | ||
| AK154807.1 | - | 19 - 20 | 2925 | 164 | ||
| AK155630.1 | - | 18 - 19 | 2699 | 164 | ||
| AK158296.1 | - | 18 - 19 | 2907 | 164 | ||
| AK162530.1 | - | 16 - 17 | 2264 | 164 | ||
| AK165710.1 | - | 3 - 4 | 872 | 164 | ||
| AK170849.1 | - | 18 - 19 | 2902 | 164 | ||
| AK171089.1 | - | 18 - 19 | 2907 | 164 | ||
| BC027417.1 | - | 4 - 5 | 1007 | 164 | ||
| BC039932.1 | - | 17 - 18 | 2442 | 164 | ||
| BC049803.1 | - | 18 - 19 | 2444 | 164 | ||
| BC063067.1 | - | 18 - 19 | 2881 | 5721396 | 164 | |
| U73200.1 | - | 18 - 19 | 2778 | 164 | ||
More Information
Set Membership: |
|

Mprip
Genomic Map