Assay Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012027.2 | NP_036157.2 | 6 - 7 | 1074 | 161 | |
| NM_201245.3 | NP_957697.1 | 6 - 7 | 1074 | 161 | ||
| GenBank mRNA | AK044512.1 | - | 6 - 7 | 824 | 161 | |
| AK154807.1 | - | 6 - 7 | 1081 | 161 | ||
| AK155630.1 | - | 6 - 7 | 855 | 161 | ||
| AK158296.1 | - | 6 - 7 | 1063 | 161 | ||
| AK162530.1 | - | 3 - 4 | 405 | 161 | ||
| AK170849.1 | - | 6 - 7 | 1058 | 161 | ||
| AK171089.1 | - | 6 - 7 | 1063 | 161 | ||
| BC039932.1 | - | 5 - 6 | 598 | 161 | ||
| BC049803.1 | - | 6 - 7 | 600 | 161 | ||
| BC063067.1 | - | 6 - 7 | 1037 | 5721396 | 161 | |
| U73200.1 | - | 6 - 7 | 934 | 161 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mprip
Genomic Map