Assay Details
Gene Symbol: Olfr1200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005227.2 | NP_001005227.2 | 1 | 952 | 169 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081525 | NM_001005227.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081525 | NM_001005227.2 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411124 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01830227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411108 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411127 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411121 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01346758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411110 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21411106 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484757 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484763 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484755 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484754 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484756 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484762 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_2876794 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2876794 | - | - |
