Assay Details
| Gene Symbol: Mrpl47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029017.2 | NP_083293.1 | 1 - 2 | 116 | 160 | |
| GenBank mRNA | BC029173.1 | - | 96 | 160 | ||
| BC038065.1 | - | 1 - 2 | 87 | 160 | ||
| BC087953.1 | - | 1 - 2 | 110 | 30119240 | 160 | |
| BY013327.1 | - | 1 - 2 | 116 | 160 | ||
| CX122942.1 | - | 1 - 2 | 95 | 160 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl47
Genomic Map