Assay Details
| Gene Symbol: Rpl18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029751.4 | NP_084027.1 | 2 - 3 | 264 | 149 | |
| GenBank mRNA | AK010983.1 | - | 2 - 3 | 235 | 149 | |
| AK146543.1 | - | 2 - 3 | 234 | 149 | ||
| AK150820.1 | - | 2 - 3 | 233 | 149 | ||
| AK152611.1 | - | 2 - 3 | 233 | 149 | ||
| AK161942.1 | - | 2 - 3 | 234 | 149 | ||
| AK162047.1 | - | 2 - 3 | 235 | 149 | ||
| AK168258.1 | - | 2 - 3 | 234 | 149 | ||
| BC037146.1 | - | 2 - 3 | 276 | 5011713 | 149 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl18a
Genomic Map