Assay Details
| Gene Symbol: Senp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163571.1 | NP_001157043.1 | 10 - 11 | 1702 | 155 | |
| NM_030702.4 | NP_109627.3 | 10 - 11 | 1914 | 155 | ||
| GenBank mRNA | AF194031.1 | - | 9 - 10 | 1595 | 155 | |
| AK045071.1 | - | 10 - 11 | 1791 | 155 | ||
| AK088608.1 | - | 10 - 11 | 1703 | 155 | ||
| AY008764.1 | - | 10 - 11 | 1646 | 155 | ||
| BC037014.1 | - | 10 - 11 | 1680 | 5344274 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896513 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070054 | BC037014.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070054 | AK045071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070054 | AY008764.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070054 | NM_030702.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070054 | NM_001163571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945701 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75433 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155226 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986386 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070054 | BC037014.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070054 | AK045071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070054 | AY008764.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070054 | NM_030702.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070054 | NM_001163571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293926 | BC037014.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293926 | AK045071.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293926 | AK088608.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293926 | AY008764.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293926 | NM_030702.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896512 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155226 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02355971 | - | - |
| Whole Mouse Genome G4122A | A_52_P574398 | BC037014.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P574398 | AK045071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P574398 | AK088608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P574398 | AY008764.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P574398 | NM_030702.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293926 | BC037014.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293926 | AK045071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293926 | AK088608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293926 | AY008764.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293926 | NM_030702.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113549 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09341747 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09288050 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09611462 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09649327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09341748 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09611463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09660586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09390341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09660589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09649327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09390340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896515 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387564 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387562 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387560 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387561 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387565 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448866_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387559 | - | - |
| Mouse Genome 430 2.0 Array | 1448866_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448866_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80886_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80886_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100027992.mm.1 | NM_001163571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027992.mm.1 | NM_001163571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027992.mm.1 | NM_030702.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027992.mm.1 | NM_030702.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027992.mm.1 | TC1100003058.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027990.mm.1 | NM_001163571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027990.mm.1 | NM_001163571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027990.mm.1 | NM_030702.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027990.mm.1 | NM_030702.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027990.mm.1 | TC1100003058.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027991.mm.1 | NM_001163571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027991.mm.1 | NM_001163571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027991.mm.1 | NM_030702.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027991.mm.1 | NM_030702.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027991.mm.1 | TC1100003058.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448866_PM_at | - | - |
| Mouse Expression Set 430 | 1448866_at | - | - |
| Murine Genome U74v2 Set | 104287_at | - | - |
| Murine Genome U74v2 Set | 104287_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448866_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448866_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448866_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448866_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 500478 | - | - |
| ABI Mouse Genome Survey Microarray | 748330 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4200082 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4200082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200082 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225635 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225635 | - | - |
| mouse-6 v1.1 (Illumina) | scl40024.13.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200082 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40024.13.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200082 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225635 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225635 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40024.13.1_29-S | - | - |

Senp3
Genomic Map