Assay Details
| Gene Symbol: Senp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163571.1 | NP_001157043.1 | 3 - 4 | 1043 | 149 | |
| NM_030702.4 | NP_109627.3 | 3 - 4 | 1255 | 149 | ||
| GenBank mRNA | AF194031.1 | - | 2 - 3 | 936 | 149 | |
| AK045071.1 | - | 3 - 4 | 1132 | 149 | ||
| AK088608.1 | - | 3 - 4 | 1044 | 149 | ||
| AY008764.1 | - | 3 - 4 | 987 | 149 | ||
| BC037014.1 | - | 3 - 4 | 1021 | 5344274 | 149 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Senp3
Genomic Map