Assay Details
| Gene Symbol: Senp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163571.1 | NP_001157043.1 | 7 - 8 | 1430 | 164 | |
| NM_030702.4 | NP_109627.3 | 7 - 8 | 1642 | 164 | ||
| GenBank mRNA | AF194031.1 | - | 6 - 7 | 1323 | 164 | |
| AK045071.1 | - | 7 - 8 | 1519 | 164 | ||
| AY008764.1 | - | 7 - 8 | 1374 | 164 | ||
| BC037014.1 | - | 7 - 8 | 1408 | 5344274 | 164 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Senp3
Genomic Map