Assay Details
Gene Symbol: Krt78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212487.4 | NP_997652.4 | 5 - 6 | 970 | 163 | |
GenBank mRNA | AY651827.1 | - | 5 - 6 | 795 | 163 | |
BC119600.1 | - | 5 - 6 | 907 | 163 | ||
BC119601.1 | - | 5 - 6 | 907 | 163 | ||
BC127241.1 | - | 5 - 6 | 907 | 163 | ||
BC127618.1 | - | 5 - 6 | 907 | 163 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054563 | AK146611.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054563 | NM_212487.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054563 | AK009535.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P311168 | AK146611.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P311168 | NM_212487.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P311168 | AK009535.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P311168 | AK146611.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P311168 | NM_212487.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P311168 | AK009535.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594278 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355351 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532860 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664854 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633628 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108755 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054563 | AK146611.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054563 | NM_212487.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054563 | AK009535.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P311168 | AK146611.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P311168 | NM_212487.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P311168 | AK009535.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P311168 | AK146611.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P311168 | NM_212487.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P311168 | AK009535.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495077 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495083 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495087 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495090 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495076 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495085 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30495112 | - | - |
Whole Mouse Genome G4122A | A_52_P311168 | AK146611.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P311168 | NM_212487.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P311168 | AK009535.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P311168 | AK146611.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P311168 | NM_212487.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P311168 | AK009535.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121080 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137762 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12134192 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12134208 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12134155 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12134195 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12134202 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12134168 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12134169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30494995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30495085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495083 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12425200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495057 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495064 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495076 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12134166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11974334 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495066 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495114 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495033 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495090 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12134192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495070 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495073 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12134206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12349532 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495058 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495077 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495094 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495071 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495069 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495091 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495116 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495061 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12134188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495059 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495086 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12134177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495074 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12134205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495029 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30495104 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10432909 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432913 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432910 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432915 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432911 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432914 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432917 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432908 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432912 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10432916 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438849_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10432907 | - | - |
Mouse Genome 430 2.0 Array | 1438849_at | - | - |
Mouse Genome 430 2.0 Array | 1438849_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 332131_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 332131_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500018813.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018813.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018813.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018817.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018817.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018817.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018818.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018818.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018818.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018815.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018815.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018815.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018819.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018819.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018819.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018809.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018809.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018809.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018811.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018811.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018811.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018810.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018810.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018810.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018816.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018816.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018816.mm.1 | TC1500002280.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500018812.mm.1 | NM_212487.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018812.mm.1 | NM_212487.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500018812.mm.1 | TC1500002280.mm.1 | MTA TC |
Mouse Expression Set 430 | 1438849_at | - | - |
Mouse Expression Set 430 | 1438849_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438849_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438849_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2489801 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2941239 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2941239 | - | - |
