Assay Details
Gene Symbol: Krt39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213730.2 | NP_998895.2 | 6 - 7 | 1204 | 133 | |
GenBank mRNA | BC119541.1 | - | 6 - 7 | 1204 | 133 | |
BC128563.1 | - | 6 - 7 | 1199 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2140007 | BC119541.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2140007 | BC128563.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2140007 | NM_213730.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341890 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680292 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213523 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675246 | AK003807.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675246 | NM_008412.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18777 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121251 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2140007 | BC119541.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2140007 | BC128563.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2140007 | NM_213730.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062922 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062912 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062918 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062883 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062924 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062916 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062900 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062915 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062901 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062914 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062920 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062921 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062908 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062887 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062897 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062879 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121251 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121251 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09514984 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09514988 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09514995 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09331961 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09514994 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09514993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09367139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09186228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09233310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062861 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09367143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062897 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09514022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09287669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062885 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062867 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062810 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062869 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062908 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09636466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062902 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09480911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09287665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09662177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062848 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09575652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09153567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062833 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062879 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062821 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062827 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09599319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062871 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062822 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09041438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09188208 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062893 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09233294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062891 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062819 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062889 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062936 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062802 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062826 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09662184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09662182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062883 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062873 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390876 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390871 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390874 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390870 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390875 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390872 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390873 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437440_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390869 | - | - |
Mouse Genome 430 2.0 Array | 1437440_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437440_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100034242.mm.1 | NM_213730.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034242.mm.1 | NM_213730.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034242.mm.1 | TC1100003710.mm.1 | MTA TC |
Mouse Expression Set 430 | 1437440_at | - | - |
Mouse Expression Set 430 | 1437440_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437440_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437440_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2699061 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2699061 | - | - |
