Assay Details
| Gene Symbol: Spag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007463.1 | NP_001007464.1 | 4 - 5 | 1056 | 165 | |
| NM_001290462.1 | NP_001277391.1 | 4 - 5 | 1056 | 165 | ||
| GenBank mRNA | AK132968.1 | - | 4 - 5 | 1057 | 165 | |
| AY769082.1 | - | 4 - 5 | 1056 | 165 | ||
| BC150868.1 | - | 4 - 5 | 1098 | 9087779 | 165 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag8
Genomic Map