Assay Details
| Gene Symbol: Airn | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035282.1 | - | 1 | 1869 | 109 | ||
| AU015441.1 | - | LIT1853 Air |
1 | 291 | 109 | ||
| BG070272.2 | - | 1 | 291 | 109 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | insulin-like growth factor 2 receptor | Igf2r |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
-48.3kb | Anti-sense | Chr. 17 - 12682406 - 12769706 on NCBI Build GRCm38 |
| Downstream | MAS1 oncogene | Mas1 |
Mas-1, Mgra |
20.5kb | Anti-sense | Chr. 17 - 12838841 - 12868145 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Airn
Genomic Map