Assay Details
Gene Symbol: Slc10a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010834.2 | NP_001010834.1 | 1 | 1280 | 145 | |
GenBank mRNA | AK143900.1 | - | 1 | 1281 | 145 | |
AY825925.1 | - | 1 | 1222 | 145 | ||
BC141377.1 | - | 1 | 1257 | 9055990 | 145 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044232 | BC141377.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044232 | AK143900.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044232 | AY825925.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044232 | NM_001010834.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495665 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417952 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171024 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122537 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044232 | BC141377.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044232 | AK143900.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044232 | AY825925.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044232 | NM_001010834.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942136 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942119 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942134 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942132 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21942138 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02999673 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02979787 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02999689 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02999674 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02918549 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02999687 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02999686 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02918548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21942072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942119 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02669991 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942121 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942103 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02901520 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02670000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942100 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02670003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02919337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02731870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02999672 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02999673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942105 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02999667 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02919331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942096 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942116 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942114 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942095 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942108 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02968139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21942111 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10497299 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10497297 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10497298 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10497296 | - | - |
Affymetrix Mouse Clariom S Array | PSR0300013077.mm.1 | NM_001010834.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300013077.mm.1 | NM_001010834.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300013077.mm.1 | TC0300001644.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300013076.mm.1 | NM_001010834.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300013076.mm.1 | NM_001010834.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300013076.mm.1 | TC0300001644.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1249970 | - | - |
