Assay Details
| Gene Symbol: Olfr988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011534.1 | NP_001011534.1 | 1 | 273 | 138 | |
| GenBank mRNA | BC150840.1 | - | 1 | 347 | 9087751 | 138 |
More Information
Set Membership: |
|

Olfr988
Genomic Map