Assay Details
Gene Symbol: Olfr1042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011777.2 | NP_001011777.1 | 1 | 223 | 159 | |
GenBank mRNA | BC150927.1 | - | 1 | 223 | 9087838 | 159 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165083 | NM_001011777.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165083 | BC150927.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305786 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165083 | NM_001011777.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165083 | BC150927.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01160298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01367970 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400961 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400970 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400955 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400972 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484562 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484561 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161343 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3161343 | - | - |
mouse-6 v1.1 (Illumina) | GI_58801329-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3161343 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161343 | - | - |
