Assay Details
| Gene Symbol: Olfr543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011782.2 | NP_001011782.2 | 1 | 988 | 82 | |
| GenBank mRNA | BC147342.1 | - | 1 | 988 | 9056415 | 82 |
| BC147343.1 | - | 1 | 988 | 9056393 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057826 | BC147343.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057826 | NM_001011782.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057826 | BC147342.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057826 | BC147343.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057826 | NM_001011782.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057826 | BC147342.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25378999 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260524 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260542 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06230088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06260530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06445592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06260540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25378986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06352552 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566137 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566136 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700032627.mm.1 | NM_001011782.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032627.mm.1 | NM_001011782.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032627.mm.1 | TC0700003859.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 584034 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161428 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161428 | - | - |
| mouse-6 v1.1 (Illumina) | GI_58801333-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161428 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161428 | - | - |

Olfr543
Genomic Map