Assay Details
Gene Symbol: Cyp4a31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252539.1 | NP_001239468.1 | 2 - 3 | 397 | 99 | |
GenBank mRNA | BC013476.1 | - | 2 - 3 | 348 | 4237837 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0400008973.mm.1 | NM_010011.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008973.mm.1 | NM_010011.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008973.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008974.mm.1 | NM_010011.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008974.mm.1 | NM_010011.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008974.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008977.mm.1 | NM_010011.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008977.mm.1 | NM_010011.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008977.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009014.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009014.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009014.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009014.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009014.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008982.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008972.mm.1 | NM_010011.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008972.mm.1 | NM_010011.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008972.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008975.mm.1 | NM_010011.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008975.mm.1 | NM_010011.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008975.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008979.mm.1 | NM_010011.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008979.mm.1 | NM_010011.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008979.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009008.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009008.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009008.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009008.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009008.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009019.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009019.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009019.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009019.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009019.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009012.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009012.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009012.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009012.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009012.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009017.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009017.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009017.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009017.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009017.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009006.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009006.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009006.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009006.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009006.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009010.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009010.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009010.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009010.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009010.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008976.mm.1 | NM_010011.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008976.mm.1 | NM_010011.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008976.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009020.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009020.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009020.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009020.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009020.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008983.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009015.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009015.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009015.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009015.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009015.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009018.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009018.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009018.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009018.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009018.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009022.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009022.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009022.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009022.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009022.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400008980.mm.1 | NM_010011.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008980.mm.1 | NM_010011.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400008980.mm.1 | TC0400004187.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009013.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009013.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009013.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009013.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009013.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009016.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009016.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009016.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009016.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009016.mm.1 | TC0400001229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400009023.mm.1 | NM_001252539.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009023.mm.1 | NM_001252539.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009023.mm.1 | NM_201640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009023.mm.1 | NM_201640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009023.mm.1 | TC0400001229.mm.1 | MTA TC |
