Assay Details
| Gene Symbol: Rnase12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011875.2 | NP_001011875.1 | 1 | 404 | 65 | |
| GenBank mRNA | BC146014.1 | - | 1 | 404 | 40131317 | 65 |
| BC146016.1 | - | 1 | 404 | 40131319 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnase12
Genomic Map