Assay Details
Gene Symbol: Olfr229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146613.1 | NP_666824.1 | 1 | 50 | 125 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993319 | NM_146613.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123038 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993319 | NM_146613.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P415047 | NM_146613.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440449 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440484 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440491 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440503 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440507 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440466 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440479 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440500 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440504 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440448 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440483 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440485 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440505 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440446 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440454 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440482 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440477 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440468 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123038 | - | - |
Whole Mouse Genome G4122A | A_52_P415047 | NM_146613.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129899 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123038 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07909248 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07939669 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07939659 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07722871 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07722895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07722895 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440449 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07544066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07520163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440455 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440452 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440505 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440514 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07939649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440491 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440503 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440511 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10584517 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10584516 | - | - |
Affymetrix Mouse Clariom S Array | PSR0900002914.mm.1 | NM_146613.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900002914.mm.1 | NM_146613.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900002914.mm.1 | TC0900000441.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 101570113 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246942 | - | - |
mouse-6 v1.1 (Illumina) | scl0258606.1_164-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101570113 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101570113 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3108633 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3034914 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3108633 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246942 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3034914 | - | - |
