Assay Details
| Gene Symbol: Spata31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030047.2 | NP_084323.2 | 3 | 3021 | 99 | |
| GenBank mRNA | AK133249.1 | - | 3 | 3022 | 99 | |
| AY842249.1 | - | 3 | 2962 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122828 | AK133249.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122828 | AY842249.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122828 | NM_030047.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155936 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117112 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101970 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122828 | AK133249.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122828 | AY842249.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122828 | NM_030047.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101970 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117112 | - | - |
| Whole Mouse Genome G4122A | A_52_P1091436 | AK133249.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1091436 | NM_030047.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1091436 | AK019634.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118070 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101970 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10669447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11032317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10976911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10828934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11032319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10719756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10669438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10669422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10976912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10722921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10669425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101129 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10405840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405839 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10405838 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300005188.mm.1 | NM_030047.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005188.mm.1 | NM_030047.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005188.mm.1 | TC1300000784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300005190.mm.1 | NM_030047.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005190.mm.1 | NM_030047.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005190.mm.1 | TC1300000784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300005189.mm.1 | NM_030047.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005189.mm.1 | NM_030047.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005189.mm.1 | TC1300000784.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 901717 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624729 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2624729 | - | - |

Spata31
Genomic Map