Assay Details
Gene Symbol: Defb39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183038.2 | NP_898859.2 | 2 | 149 | 93 | |
GenBank mRNA | AJ575425.1 | - | 2 | 93 | ||
AJ578472.1 | - | 2 | 149 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613853 | NM_183038.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613853 | AJ575425.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613853 | AJ578472.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613853 | NM_183038.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613853 | AJ575425.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613853 | AJ578472.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712844 | AK030285.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56772 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967653 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171598 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613853 | NM_183038.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613853 | AJ575425.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613853 | AJ578472.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613853 | NM_183038.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613853 | AJ575425.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613853 | AJ578472.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701884 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701874 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701887 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25701880 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01688580 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171598 | - | - |
Whole Mouse Genome G4122A | A_52_P613853 | NM_183038.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P613853 | AJ575425.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P613853 | AJ578472.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P613853 | NM_183038.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P613853 | AJ575425.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P613853 | AJ578472.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111424 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171598 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01688580 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07335766 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07335771 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07276205 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07335773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25701901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07225919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06947334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701892 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701869 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07387126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07335766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701874 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701872 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07367900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701889 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701879 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07065214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701885 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25701868 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10577350 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577351 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10577349 | - | - |
Affymetrix Mouse Clariom S Array | PSR0800014399.mm.1 | NM_183038.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800014399.mm.1 | NM_183038.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800014399.mm.1 | TC0800001805.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800014400.mm.1 | NM_183038.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800014400.mm.1 | NM_183038.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800014400.mm.1 | TC0800001805.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800014401.mm.1 | NM_183038.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800014401.mm.1 | NM_183038.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800014401.mm.1 | TC0800001805.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2480168 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2480168 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002480168 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2774388 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2774388 | - | - |
mouse-6 v1.1 (Illumina) | scl0360214.1_45-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2480168 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0360214.1_45-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480168 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2774388 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2774388 | - | - |
