Assay Details
| Gene Symbol: Cxcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030712.4 | NP_109637.3 | 2 | 1714 | 113 | |
| GenBank mRNA | AF305709.1 | - | 2 | 1719 | 113 | |
| AK052901.1 | - | 2 | 1712 | 113 | ||
| AK137703.1 | - | 2 | 1693 | 113 | ||
| AK153914.1 | - | 2 | 1713 | 113 | ||
| BC096491.1 | - | 2 | 1734 | 30748222 | 113 | |
| BQ562712.1 | - | 1 | 113 | |||
| JC209414.1 | - | 2 | 1714 | 113 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chemotaxis
inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
chemokine-mediated signaling pathway
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
coreceptor activity
C-X-C chemokine receptor activity
C-X-C chemokine binding
chemotaxis
inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
chemokine-mediated signaling pathway
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
coreceptor activity
C-X-C chemokine receptor activity
C-X-C chemokine binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165199 | AK137703.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165199 | AK153914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165199 | BC096491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165199 | BQ562712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165199 | NM_030712.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962209 | AK137703.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962209 | AK153914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962209 | BC096491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962209 | BQ562712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962209 | NM_030712.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125491 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103608 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152309 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165199 | AK137703.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165199 | AK153914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165199 | BC096491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165199 | BQ562712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165199 | NM_030712.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962209 | AK137703.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962209 | AK153914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962209 | BC096491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962209 | BQ562712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962209 | NM_030712.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P412027 | AK137703.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P412027 | AK153914.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P412027 | BC096491.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P412027 | BQ562712.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P412027 | NM_030712.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26896979 | - | - |
| Whole Mouse Genome G4122A | A_51_P412027 | AK137703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P412027 | AK153914.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P412027 | BC096491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P412027 | BQ562712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P412027 | NM_030712.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121864 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08055756 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08055765 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08055762 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08055750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26896987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08055758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08055764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07797103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08198755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07596709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08055762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07723793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07723792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07719220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26896978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07723795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07519618 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10590625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590624 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422812_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425832_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10590623 | - | - |
| Mouse Genome 430 2.0 Array | 1425832_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422812_at | - | - |
| Mouse Genome 430 2.0 Array | 1422812_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425832_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80901_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80901_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900014060.mm.1 | NM_030712.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900014060.mm.1 | NM_030712.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900014060.mm.1 | TC0900001672.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900014059.mm.1 | TC0900001672.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900014058.mm.1 | NM_030712.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900014058.mm.1 | NM_030712.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900014058.mm.1 | TC0900001672.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425832_PM_a_at | - | - |
| Mouse Expression Set 430 | 1422812_at | - | - |
| Mouse Expression Set 430 | 1425832_a_at | - | - |
| Mouse Expression Set 430 | 1422812_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425832_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422812_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422812_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425832_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422812_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425832_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422812_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425832_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 801088 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3190440 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190440 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221846 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221846 | - | - |
| mouse-6 v1.1 (Illumina) | scl36285.2_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190440 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36285.2_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190440 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221846 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221846 | - | - |

Cxcr6
Genomic Map