Assay Details
| Gene Symbol: Klhl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182782.2 | NP_877583.1 | 2 | 2164 | 71 | |
| GenBank mRNA | AK157879.1 | - | 2 | 1213 | 71 | |
| BC027373.1 | - | 2 | 2166 | 4948451 | 71 | |
| BP767188.1 | - | 1 | 71 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Klhl25
Genomic Map