Assay Details
| Gene Symbol: Ndufs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010888.2 | NP_035018.1 | 3 | 275 | 102 | |
| NR_033306.1 | 3 | 336 | 102 | |||
| GenBank mRNA | AK147147.1 | - | 3 | 305 | 102 | |
| BC059730.1 | - | 3 | 248 | 102 | ||
| BC076501.1 | - | 3 | 306 | 102 | ||
| BC086933.1 | - | 3 | 248 | 6760537 | 102 | |
| BG081040.2 | - | 3 | 265 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid metabolic process
transport
muscle contraction
multicellular organism aging
respiratory electron transport chain
multicellular organism growth
oxidation-reduction process
reproductive system development
mitochondrion morphogenesis
cardiovascular system development
molecular_function
fatty acid metabolic process
transport
muscle contraction
multicellular organism aging
respiratory electron transport chain
multicellular organism growth
oxidation-reduction process
reproductive system development
mitochondrion morphogenesis
cardiovascular system development
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139147 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139132 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964867 | NM_010888.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964867 | NR_033306.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964867 | BC076501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964867 | BC086933.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9305 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155972 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964867 | NM_010888.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964867 | NR_033306.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964867 | BC076501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964867 | BC086933.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P375034 | NM_010888.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P375034 | NR_033306.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P375034 | AK147147.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P375034 | BC059730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P375034 | BC076501.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335077 | NM_010888.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335077 | NR_033306.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335077 | AK147147.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335077 | BC059730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335077 | BC076501.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139119 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07917920 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155972 | - | - |
| Whole Mouse Genome G4122A | A_52_P685828 | NM_010888.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685828 | NR_033306.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685828 | AK147147.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685828 | BC059730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685828 | BC076501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375034 | NM_010888.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375034 | NR_033306.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375034 | AK147147.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375034 | BC059730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375034 | BC076501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335077 | NM_010888.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335077 | NR_033306.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335077 | AK147147.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335077 | BC059730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335077 | BC076501.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155972 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10559445 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10559447 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10559457 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969731 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10559449 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10467629 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969726 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969722 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10559453 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139147 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10878537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10766848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10766854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10559457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10766883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10766861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10766892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10766888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10614353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10766859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139109 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10410509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410511 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433603_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10410508 | - | - |
| Mouse Genome 430 2.0 Array | 1433603_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433603_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 407785_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 407785_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300015053.mm.1 | NM_010888.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300015053.mm.1 | NM_010888.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300015053.mm.1 | NR_033306.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300015053.mm.1 | NR_033306.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300015053.mm.1 | TC1300002299.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433603_at | - | - |
| Mouse Expression Set 430 | 1433603_PM_at | - | - |
| Murine Genome U74v2 Set | 96291_PM_f_at | - | - |
| Murine Genome U74v2 Set | 96291_f_at | - | - |
| Murine Genome U74v2 Set | 96292_PM_r_at | - | - |
| Murine Genome U74v2 Set | 96292_r_at | - | - |
| Murine Genome U74v2 Set | 171129_r_at | - | - |
| Murine Genome U74v2 Set | 171129_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433603_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433603_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433603_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433603_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 896596 | - | - |
| ABI Mouse Genome Survey Microarray | 900173 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2842572 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2842572 | - | - |

Ndufs6
Genomic Map