Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000008173.mm.1 |
NM_001004164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008173.mm.1 |
NM_001004164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008173.mm.1 |
TC1000001079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008174.mm.1 |
NM_001004164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008174.mm.1 |
NM_001004164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008174.mm.1 |
TC1000001079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008175.mm.1 |
NM_001004164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008175.mm.1 |
NM_001004164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008175.mm.1 |
TC1000001079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008172.mm.1 |
NM_001004164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008172.mm.1 |
NM_001004164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008172.mm.1 |
TC1000001079.mm.1 |
MTA TC |