Assay Details
| Gene Symbol: Gnptab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004164.2 | NP_001004164.2 | 7 - 8 | 778 | 118 | |
| GenBank mRNA | AK140867.1 | - | 6 - 7 | 882 | 118 | |
| AK173132.1 | - | 7 - 8 | 778 | 118 | ||
| BC060638.1 | - | 8 - 9 | 886 | 6849405 | 118 | |
More Information
Set Membership: |
|

Gnptab
Genomic Map