Assay Details
| Gene Symbol: Gnptab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004164.2 | NP_001004164.2 | 3 - 4 | 331 | 69 | |
| GenBank mRNA | AK140867.1 | - | 2 - 3 | 435 | 69 | |
| AK173132.1 | - | 3 - 4 | 331 | 69 | ||
| BC060638.1 | - | 4 - 5 | 439 | 6849405 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060638.1 transcripts.
Set Membership: |
|

Gnptab
Genomic Map