Assay Details
| Gene Symbol: Ces1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013764.2 | NP_001013786.2 | 11 - 12 | 1392 | 94 | |
| GenBank mRNA | BC089371.1 | - | 11 - 12 | 1392 | 30604811 | 94 |
More Information
Set Membership: |
|

Ces1a
Genomic Map