Assay Details
Gene Symbol: Ces1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013764.2 | NP_001013786.2 | 13 - 14 | 1601 | 103 | |
GenBank mRNA | BC089371.1 | - | 13 - 14 | 1601 | 30604811 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952577 | BC089371.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945871 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838704 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640298 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462389 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258939 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399616 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230871 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816754 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779159 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722663 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831255 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387583 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616906 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741567 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811455 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67272 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94492 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952577 | BC089371.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069549 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069541 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069561 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069545 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069559 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069563 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069547 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069557 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069553 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069548 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069554 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069550 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069555 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069542 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069560 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26069556 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01798779 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06599141 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06599110 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06599110 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01798779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07147968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06902407 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07330996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07128927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06937656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06707844 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07366422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07011759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06721921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069348 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069547 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07088641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07343314 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07330995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07128932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26069548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06721920 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10580607 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580600 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580595 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580602 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580596 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580599 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580594 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580605 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580604 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580603 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580597 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580598 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580601 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580606 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10580593 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1237261 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2784942 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2784942 | - | - |
