Assay Details
| Gene Symbol: Ces1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013764.2 | NP_001013786.2 | 1 - 2 | 140 | 69 | |
| GenBank mRNA | BC089371.1 | - | 1 - 2 | 140 | 30604811 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Ces1a
Genomic Map