Assay Details
| Gene Symbol: Ces1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013764.2 | NP_001013786.2 | 8 - 9 | 1024 | 84 | |
| GenBank mRNA | BC089371.1 | - | 8 - 9 | 1024 | 30604811 | 84 |
More Information
Set Membership: |
|

Ces1a
Genomic Map