Assay Details
| Gene Symbol: Naa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033191.2 | NP_001028363.1 | 1 - 2 | 708 | 77 | |
| GenBank mRNA | AK133248.1 | - | 1 - 2 | 716 | 77 | |
| AK135863.1 | - | 1 - 2 | 710 | 77 | ||
| BC139104.1 | - | 1 - 2 | 709 | 8862126 | 77 | |
| BC145776.1 | - | 1 - 2 | 709 | 40131079 | 77 | |
| EU192141.1 | - | 1 - 2 | 757 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
N-terminal protein amino acid acetylation
internal protein amino acid acetylation
N-terminal peptidyl-serine acetylation
N-terminal peptidyl-glutamic acid acetylation
peptide alpha-N-acetyltransferase activity
N-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups
peptide-serine-N-acetyltransferase activity
peptide-glutamate-N-acetyltransferase activity
N-terminal protein amino acid acetylation
internal protein amino acid acetylation
N-terminal peptidyl-serine acetylation
N-terminal peptidyl-glutamic acid acetylation
peptide alpha-N-acetyltransferase activity
N-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups
peptide-serine-N-acetyltransferase activity
peptide-glutamate-N-acetyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23912413 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23912412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23912410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23912411 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P05456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571104 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48619 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P05456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912398 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05664189 | - | - |
| Whole Mouse Genome G4122A | A_51_P282297 | BC139104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282297 | BC145776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282297 | EU192141.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282297 | AK133248.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282297 | AK135863.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05664189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23912412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23912411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23912410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23912413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04669213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04946954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04412284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04310153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04743597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04536857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04536855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04946964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04786400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04536858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04262652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04786415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04832363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04367880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04946957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04946970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04946969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04786422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04808996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04946960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04283611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04566005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912387 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10531554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531555 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437786_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458895_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531553 | - | - |
| Mouse Genome 430 2.0 Array | 1437786_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458895_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437786_at | - | - |
| Mouse Genome 430 2.0 Array | 1458895_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 97243_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 97243_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500024631.mm.1 | TC0500002802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500024631.mm.1 | NM_001033191.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500024631.mm.1 | NM_001033191.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500024629.mm.1 | TC0500002802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500024629.mm.1 | NM_001033191.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500024629.mm.1 | NM_001033191.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500024630.mm.1 | TC0500002802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500024630.mm.1 | NM_001033191.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500024630.mm.1 | NM_001033191.2 | Refseq |
| Mouse Expression Set 430 | 1437786_at | - | - |
| Mouse Expression Set 430 | 1437786_PM_at | - | - |
| Mouse Expression Set 430 | 1458895_PM_at | - | - |
| Mouse Expression Set 430 | 1458895_at | - | - |
| Murine Genome U74v2 Set | 160945_at | - | - |
| Murine Genome U74v2 Set | 160945_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437786_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437786_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437786_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437786_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458895_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458895_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2944168 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2944168 | - | - |

Naa11
Genomic Map