Assay Details
| Gene Symbol: Rimbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033338.3 | NP_001028510.2 | 1 | 5717 | 135 | |
| GenBank mRNA | AK133200.1 | - | 1 | 4868 | 135 | |
| BG075083.2 | - | 1 | 135 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572556 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572551 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572555 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572547 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572550 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572545 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128442 | NM_001033338.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128442 | AK133200.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128442 | BG075083.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556133 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31906 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128442 | NM_001033338.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128442 | AK133200.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128442 | BG075083.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572546 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08520160 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08520160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572551 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572550 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572545 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572556 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572547 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12837842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12694504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12795551 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10433972 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443875_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433971 | - | - |
| Mouse Genome 430 2.0 Array | 1443875_at | - | - |
| Mouse Genome 430 2.0 Array | 1443875_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 239731_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 239731_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600001705.mm.1 | NM_001033338.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001705.mm.1 | NM_001033338.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001705.mm.1 | TC1600000211.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600001702.mm.1 | NM_001033338.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001702.mm.1 | NM_001033338.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001702.mm.1 | TC1600000211.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600001703.mm.1 | NM_001033338.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001703.mm.1 | NM_001033338.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001703.mm.1 | TC1600000211.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600001704.mm.1 | NM_001033338.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001704.mm.1 | NM_001033338.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001704.mm.1 | TC1600000211.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1443875_at | - | - |
| Mouse Expression Set 430 | 1443875_PM_at | - | - |
| Murine Genome U74v2 Set | 168025_at | - | - |
| Murine Genome U74v2 Set | 168025_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443875_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443875_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2934163 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2934163 | - | - |

Rimbp3
Genomic Map