| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900018010.mm.1 |
NM_001033469.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018010.mm.1 |
NM_001033469.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018010.mm.1 |
TC0900002166.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900018008.mm.1 |
NM_001033469.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018008.mm.1 |
NM_001033469.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018008.mm.1 |
TC0900002166.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900018009.mm.1 |
NM_001033469.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018009.mm.1 |
NM_001033469.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018009.mm.1 |
TC0900002166.mm.1 |
MTA TC |