Assay Details
| Gene Symbol: Foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033469.2 | NP_001028641.1 | 2 - 3 | 189 | 91 | |
| GenBank mRNA | AK139562.1 | - | 2 - 3 | 189 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Foxr1
Genomic Map