Assay Details
| Gene Symbol: Foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033469.2 | NP_001028641.1 | 4 - 5 | 593 | 110 | |
| GenBank mRNA | AK139562.1 | - | 4 - 5 | 593 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
anatomical structure morphogenesis
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
anatomical structure morphogenesis
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467611 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467613 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467610 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005883 | NM_001033469.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005883 | AK139562.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302168 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130972 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005883 | NM_001033469.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005883 | AK139562.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06769472 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06769472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467610 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467611 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08016228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07580924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07892936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07892935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08095848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07981146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08016237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08016239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08041292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07925866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07728163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07698674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07710776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08016243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07925860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467603 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592877 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592870 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900018005.mm.1 | NM_001033469.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018005.mm.1 | NM_001033469.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018005.mm.1 | TC0900002166.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900018004.mm.1 | NM_001033469.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018004.mm.1 | NM_001033469.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018004.mm.1 | TC0900002166.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162291 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3162291 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85701637-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3162291 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162291 | - | - |

Foxr1
Genomic Map