| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300000984.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000984.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000984.mm.1 |
TC0300000185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000985.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000985.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000985.mm.1 |
TC0300000185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000987.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000987.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000987.mm.1 |
TC0300000185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000983.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000983.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000983.mm.1 |
TC0300000185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000988.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000988.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000988.mm.1 |
TC0300000185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000986.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000986.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000986.mm.1 |
TC0300000185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000982.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000982.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000982.mm.1 |
TC0300000185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000980.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000980.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000980.mm.1 |
TC0300000185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000981.mm.1 |
NM_001033479.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000981.mm.1 |
NM_001033479.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000981.mm.1 |
TC0300000185.mm.1 |
MTA TC |